Polypropylene Swing-Out 4-in. Paddle Assembly, 5/16 x 13-13/16 in.
Copy product name
Copy catalog number
Copy catalog number and product name
Copy link
Cole-Parmer™
Polypropylene Swing-Out 4-in. Paddle Assembly, 5/16 x 13-13/16 in.
Maximize your mixing
Have Questions?
Catalog number CP0636760
also known as CP-CP0636760-EACH
Price (KRW)
-
Contact Us
Ask our AI about this Product
Maximize your mixing with these accessories for laboratory mixers
Specifications
Product TypePaddle Assembly
Diameter (English) Shaft0.31 in.
Diameter (Metric) Shaft8 mm
Length (English)13.8125 in.
Length (English) Shaft13.8125 in.
Length (Metric) Shaft350 mm
Blade MaterialPolypropylene
MaterialPolypropylene
DescriptionPolypropylene swing-out paddle assembly; 5/16 in. dia x 13-13/16 in. L, 4 in. paddle dia
For Use With (Equipment)Laboratory Mixer
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our
Privacy Notice.