Oakton pH 6+ Handheld Meter with pH Probe
Oakton pH 6+ Handheld Meter with pH Probe
Cole-Parmer™

Oakton pH 6+ Handheld Meter with pH Probe

Oakton pH 6+ Handheld pH Meters
Have Questions?
Catalog NumberRange (pH)
CP3561322
also known as CP-CP3561322-EACH
0 to 14
Catalog number CP3561322
also known as CP-CP3561322-EACH
Price (KRW)
-
Range (pH):
0 to 14
Ask our AI about this Product
Oakton pH 6+ Handheld pH Meters

High +/-0.01 pH accuracy at an economical price

  • Convenient, compact design
  • Up to five-point push-button calibration with autobuffer recognition
  • Displays pH or mV and temperature with the press of a button
Specifications
Accuracy (mV)±0.2 or ±0.05%, whichever is greater
Accuracy (pH)±0.01
Battery Life700 hr.
Battery Type4 x AAA batteries (included)
DescriptionHandheld Meter with pH Probe
For Use With (Application)NIST and USA Buffers
Height (English)1.6875 in.
Height (Metric)4.28625 cm
Resolution (mV)0.1 or 1 mV
Temperature CompensationAutomatic
Temperature Range (English)14°F to 230°F
TypePortable
Width (English)3.375 in.
Width (Metric)8.5725 cm
Calibration PointsUp to 5
Display TypeLCD
Range (mV)-1999 to 1999
Range (pH)0 to 14
Resolution (pH)0.01, 0.1
Temperature Range (Metric)-10°C to +110°C
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

0 results displayed, search above for a specific certificate

Share catalog number, name or link

1x1 image pixel for data collection