Potassium Phosphate, Monobasic, Molecular Biology Grade, Calbiochem™, , 1kg
Potassium Phosphate, Monobasic, Molecular Biology Grade, Calbiochem™, , 1kg
Potassium Phosphate, Monobasic, Molecular Biology Grade, Calbiochem™, , 1kg
Potassium Phosphate, Monobasic, Molecular Biology Grade, Calbiochem™, , 1kg
Fisher Scientific™

Potassium Phosphate, Monobasic, Molecular Biology Grade, Calbiochem™, , 1kg

Have Questions?
보기 방식 변경buttonViewtableView
수량:
250 g
1 kg
카탈로그 번호 529568250GM
FISHERMIS-529568250GM-EACH으로도 사용됨
제품 가격(KRW)
-
수량:
250 g
대량 주문 또는 맞춤형 요청
사양
화학물질 이름 또는 재질Potassium Phosphate, Monobasic
CAS7778-77-0
권장 스토리지Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).
RTECS 번호TC6615500
수량250 g
더 보기
Ask our AI about this Product
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

문서 및 다운로드

Certificates | 증명서

시험 성적서 관련 0 개의 결과가 검색되었습니다.

카탈로그 번호, 제품명 또는 링크 공유

1x1 image pixel for data collection