No se puede solicitar en la web
El producto 60180-M177: SureSTART™ WebSeal™ Clear Alcohol Resistant EVA 96-Well Plate Sealing Mats, 8 mm Diameter, Square, Flat Base, Level 1 Everyday Analysis, 5 Pk. no se puede solicitar en su país en este momento. Póngase en contacto con nuestro servicio de asistencia técnica para conocer las posibles alternativas.
Explore otros productos de esta categoría
Microplate Sealing Mats
Microplate sealing mats, which cover the surface of the plate, protect contents from leakage, contamination, and evaporation during assay processing, incubation, or storage by using molded well caps to simultaneously seal each well. They are suitable for applications at a wide range of temperatures, including autoclaving, and storage at temperatures to -80°C. Microplate sealing mats are available for different plate types, with flat or domed well caps, and in a variety of materials and colors. For easy well access without removing the mat, choose the pre-slit and pierceable format, which can be used with a pipette or needle. Mats can be applied manually or with a storage mat applicator. Safely seal your plates and eliminate cross-contamination of samples with Thermo Scientific™ MicroMat™ CLR Silicone Mats for Well Plates. Seal 96 samples at one time and achieve maximum space saving benefits with the high-density storage format of the Thermo Scientific™ Abgene™ SmartCap Mat.
1x1 image pixel for data collection