Compatible with many prepared media formats, theeasy-to-clean, stainless steel DiluFlux Pro Dilutors are an ideal fit for any food testing laboratory.
Automated media dispensing and sample dilution
- Compact and easy-to-use
- Automated, rapid and accurate dispensing
- Available with one or two pump heads for multiple media dispensing
- Compatible with Dry-Bags and other media formats