Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: CI2Z9E .
Sequence of this protein is as follows: SIADRGSFES ASELRIQLRR THQADHVPII LVGNKADLAR CREVSVEEGR ACAVVFDCKF IETSATLQHN VAELFEGVVR QLRLRRRDSA
The protein encoded by this gene is a GTPase and member of the RAS-like GTP-binding protein family. The encoded protein is expressed in endothelial cells, where it promotes reorganization of the actin cytoskeleton and morphological changes in the cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GTP-binding protein REM 1; GTPase-regulating endothelial cell sprouting; Rad and Gem-like GTP-binding protein 1; RAS (RAD and GEM)-like GTP-binding 1
Gene Aliases: GD:REM; GES; REM; REM1
UniProt ID: (Human) O75628
Entrez Gene ID: (Human) 28954
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support