Invitrogen™

Nitrocellulose/Filter Paper Sandwich, 0.2 μm, 8.3 x 7.3 cm

Catalog number: LC2000
Have Questions?
Invitrogen™

Nitrocellulose/Filter Paper Sandwich, 0.2 μm, 8.3 x 7.3 cm

Catalog number: LC2000
Have Questions?
Catalog Number
LC2000
Unit Size
20 membrane/filter paper sandwiches
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
LC200020 membrane/filter paper sandwiches
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Nitrocellulose/Filter Paper Sandwiches, 0.2 μm, are high-quality membranes ideal for blotting proteins and nucleic acids. The 0.2-μm pore size is ideal for the transfer of low molecular weight proteins (less than 20 kDa) and nucleic acids (less than 300 bp).

See all membranes and filter papers ›

Features:
Membrane: 100% pure nitrocellulose
Filter paper thickness: 0.8 mm
Binding capacity: ∼209 μg/cm2 of proteins
Binding interactions: hydrophobic and electrostatic
Compatibility: compatible with commonly used transfer conditions and detection methods such as staining, immunodetection, chemiluminescence, fluorescence, and radiolabeling
• Supplied in a pre-cut, pre-assembled membrane/filter paper sandwich
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Length (Metric)
8.3 cm
Width (Metric)
7.3 cm
Description
Nitrocellulose Pre-Cut Blotting Membrane, 0.2μm pore size
Format
Sandwich
Material
Nitrocellulose
Pore Size
0.2 μm
Shipping Condition
Room Temperature
Dimensions (LxW)
7.3 cm x 8.3 cm
Thickness
0.8 mm
Product Line
Novex

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text