Potassium thiocyanate, 0.1N Standardized Solution, Thermo Scientific Chemicals
Potassium thiocyanate, 0.1N Standardized Solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium thiocyanate, 0.1N Standardized Solution, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
Catalog number 035602.K2
also known as 035602-K2
Price (TWD)/ Each
-
Request A Quote
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS333-20-0
IUPAC Namepotassium cyanosulfanide
Molecular FormulaCKNS
InChI KeyZNNZYHKDIALBAK-UHFFFAOYSA-M
SMILES[K+].[S-]C#N
View more
SpecificationsSpecification SheetSpecification Sheet
MatrixWater
Concentration0.1N ? 0.2%
Potassium thiocyanate, 0.1N standardized solution is used in analytical chemistry for the quantitative determination of Fe(III). In Volhard’s method, it is used to determine the concentration of chloride ion by titration. It is also used as an analyzer for direct and continuous measurement of the concentration of soluble copper in an aqueous solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only