1,5-Anhydro-D-sorbitol, 97%, Thermo Scientific Chemicals
1,5-Anhydro-D-sorbitol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,5-Anhydro-D-sorbitol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 459912500
Price (TWD)/ Each
-
Request A Quote
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
IUPAC Name(2R,3S,4R,5S)-2-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC6H12O5
InChI KeyMPCAJMNYNOGXPB-SLPGGIOYSA-N
SMILESOC[C@H]1OC[C@H](O)[C@@H](O)[C@@H]1O
Molecular Weight (g/mol)164.16
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder or crystalline powder
Melting point140°C to 145°C
Elemental Analysis>=42.14 to =<45.66 % Carbon
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only