Ertapenem sodium, 85%
Ertapenem sodium, 85%
Ertapenem sodium, 85%
Ertapenem sodium, 85%
Thermo Scientific Chemicals

Ertapenem sodium, 85%

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
4606400101g
460642500250mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 460640010
Price (TWD)
-
Request A Quote
Quantity:
1g
Request bulk or custom format
Chemical Identifiers
CAS153773-82-1
IUPAC Namesodium 3-[(2S,4S)-4-{[(4R,5S,6S)-2-carboxy-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-en-3-yl]sulfanyl}pyrrolidine-2-amido]benzoate
Molecular FormulaC22H24N3NaO7S
InChI KeyZXNAQFZBWUNWJM-HRXMHBOMSA-M
SMILES[Na+].C[C@@H](O)[C@@H]1[C@H]2[C@@H](C)C(S[C@@H]3CN[C@@H](C3)C(=O)NC3=CC=CC(=C3)C([O-])=O)=C(N2C1=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow to light brown
Infrared spectrumConforms
HPLC>=80 %
Loss on drying15 % to 19 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link