Thermo Scientific Chemicals

2-Fluorophenylacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A10596.09
10 g, Each
Thermo Scientific Chemicals

2-Fluorophenylacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A10596.09
10 g, Each
Quantity
Catalog number: A10596.09
also known as A10596-09
Price (TWD)

Chemical Identifiers

CAS
451-82-1
IUPAC Name
2-(2-fluorophenyl)acetic acid
Molecular Formula
C8H7FO2
InChI Key
RPTRFSADOICSSK-UHFFFAOYSA-N
SMILES
OC(=O)CC1=CC=CC=C1F
Form
Crystals or powder or crystalline powder or flakes
Melting Point (clear melt)
60.0-67.0?C
Assay (Silylated GC)
≥98.0%
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text