Thermo Scientific™

4-Bromophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: A15590.06
5 g, Each
Thermo Scientific™

4-Bromophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: A15590.06
5 g, Each
Quantity
Catalog number: A15590.06
also known as A15590-06
Price (TWD)

Chemical Identifiers

CAS
622-88-8
IUPAC Name
hydrogen (4-bromophenyl)hydrazine chloride
Molecular Formula
C6H8BrClN2
InChI Key
RGGOWBBBHWTTRE-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NNC1=CC=C(Br)C=C1
Form
Crystals or powder or crystalline powder or flakes
Appearance (Color)
White to cream to brown or pink
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%

Description

4-Bromophenylhydrazine hydrochloride is used as a reagent in the preparation of acylsulfonamides, acylsulfamides and 9-bromo-2-ethenyl-7,12-dihydroindolo[3,2-d][1]benzazepin-6(5H)-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromophenylhydrazine hydrochloride is used as a reagent in the preparation of acylsulfonamides, acylsulfamides and 9-bromo-2-ethenyl-7,12-dihydroindolo[3,2-d][1]benzazepin-6(5H)-one.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text