Thermo Scientific Chemicals

trans,trans-1,4-Diphenyl-1,3-butadiene, 98+%, Thermo Scientific Chemicals

Catalog number: A17531.14
25 g, Each
Thermo Scientific Chemicals

trans,trans-1,4-Diphenyl-1,3-butadiene, 98+%, Thermo Scientific Chemicals

Catalog number: A17531.14
25 g, Each
Quantity
Catalog number: A17531.14
also known as A17531-14
Price (TWD)

Chemical Identifiers

CAS
538-81-8
IUPAC Name
[(1E,3E)-4-phenylbuta-1,3-dien-1-yl]benzene
Molecular Formula
C16H14
InChI Key
JFLKFZNIIQFQBS-FNCQTZNRSA-N
SMILES
C(\C=C\C1=CC=CC=C1)=C/C1=CC=CC=C1
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
White to cream
Assay (GC)
≥98.0%
Melting Point (clear melt)
147.0-157.0?C

Description

trans,trans-1,4-Diphenyl-1,3-butadiene reacts with Rieke metal complexes of barium and strontium to prepare metal-diene reagents, which is used for the carbocyclization with dichloroalkanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans,trans-1,4-Diphenyl-1,3-butadiene reacts with Rieke metal complexes of barium and strontium to prepare metal-diene reagents, which is used for the carbocyclization with dichloroalkanes.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text