Thermo Scientific Chemicals

2-Ethylimidazole, 98%, Thermo Scientific Chemicals

Catalog number: B24090.36
500 g, Each
Thermo Scientific Chemicals

2-Ethylimidazole, 98%, Thermo Scientific Chemicals

Catalog number: B24090.36
500 g, Each
Quantity
Catalog number: B24090.36
also known as B24090-36
Price (TWD)
Have Questions?

Chemical Identifiers

CAS
1072-62-4
IUPAC Name
2-ethyl-1H-imidazole
Molecular Formula
C5H8N2
InChI Key
PQAMFDRRWURCFQ-UHFFFAOYSA-N
SMILES
CCC1=NC=CN1
Appearance (Color)
White to pale yellow
Assay (GC)
≥97.5%
Melting Point (clear melt)
81.5-87.5?C
Water Content (Karl Fischer Titration)
≤1.0%
Form
Crystals or powder or crystalline powder or flakes

Description

2-Ethylimidazole is used as internal standard in the quantification of 4-(5-)methylimidazole (4MI) in coffee by GC-MS method. It is a versatile intermediate used as a building block for active ingredients as well as in epoxy curing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylimidazole is used as internal standard in the quantification of 4-(5-)methylimidazole (4MI) in coffee by GC-MS method. It is a versatile intermediate used as a building block for active ingredients as well as in epoxy curing.

Solubility
Soluble in water (617 g/L at 20°C)

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text