Absorbance≤0.01(0.1M solution) at 260nm, 0.004 max. (0.1M solution) at 280nm
CAS7365-45-9
CAS7732-18-5
ChemAlert Storage SymbolGray
Chemical Name or MaterialHEPES Buffer
View more
Ask our AI about this Product
Commonly used buffering agent
HEPES is commonly used as a buffering agent.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our
Privacy Notice.
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
0 results displayed, search above for a specific certificate