Thermo Scientific Chemicals

6-(Trifluoromethyl)pyridine-3-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: H34033.03
1 g, Each
Thermo Scientific Chemicals

6-(Trifluoromethyl)pyridine-3-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: H34033.03
1 g, Each
Quantity
Catalog number: H34033.03
also known as H34033-03
Price (TWD)

Chemical Identifiers

CAS
959996-58-8
IUPAC Name
6-(trifluoromethyl)pyridine-3-sulfonyl chloride
Molecular Formula
C6H3ClF3NO2S
InChI Key
NSGLNIQAWVNZFB-UHFFFAOYSA-N
SMILES
FC(F)(F)C1=NC=C(C=C1)S(Cl)(=O)=O
Form
Crystals or powder or crystalline powder or fused solid
Appearance (Color)
White to cream or pale yellow
Assay (GC)
≥96.0%
Melting Point (clear melt)
41-46°C
Identification (FTIR)
Conforms

Description

6-Trifluoromethyl-3-pyridinesulfonyl Chloride is a chemical used for the preparation of N-sulfonamido polycyclic pyrazolyl compounds for treatment of cognitive disorders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Trifluoromethyl-3-pyridinesulfonyl Chloride is a chemical used for the preparation of N-sulfonamido polycyclic pyrazolyl compounds for treatment of cognitive disorders.

Notes
Store in cool place. Keep container ti
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text