(S)-N-Fmoc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 95%
(S)-N-Fmoc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 95%
(S)-N-Fmoc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 95%
(S)-N-Fmoc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 95%
Thermo Scientific Chemicals

(S)-N-Fmoc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 95%

CAS: 136030-33-6 | C25H21NO4 | 399.446 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H51975.03
also known as H51975-03
Price (TWD)
-
Request A Quote
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS136030-33-6
IUPAC Name(3S)-2-{[(9H-fluoren-9-yl)methoxy]carbonyl}-1,2,3,4-tetrahydroisoquinoline-3-carboxylate
Molecular FormulaC25H20NO4
InChI KeyLIRBCUNCXDZOOU-QHCPKHFHSA-M
SMILES[O-]C(=O)[C@@H]1CC2=CC=CC=C2CN1C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Color ChangeWhite to off-white
Optical RotationReport Value
FormPowder
Purity≥94.0%
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
X17I033Certificate of AnalysisNov 09, 2022H51975.06, H51975.03
N02I039Certificate of AnalysisApr 12, 2022H51975.06, H51975.03
K17W058Certificate of AnalysisDec 07, 2021H51975.06, H51975.03
Z08C017Certificate of AnalysisDec 07, 2021H51975.06, H51975.03
4 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection