Thermo Scientific Chemicals

2-(2-Fluorophenyl)pyrrolidine, 95%, Thermo Scientific Chemicals

Catalog number: H61699.03
1 g, Each
Thermo Scientific Chemicals

2-(2-Fluorophenyl)pyrrolidine, 95%, Thermo Scientific Chemicals

Catalog number: H61699.03
1 g, Each
Quantity
Catalog number: H61699.03
also known as H61699-03
Price (TWD)
Have Questions?

Chemical Identifiers

CAS
72216-04-7
IUPAC Name
2-(2-fluorophenyl)pyrrolidine
Molecular Formula
C10H12FN
InChI Key
NOJCVVKIBLHAGW-UHFFFAOYNA-N
SMILES
FC1=CC=CC=C1C1CCCN1
Assay (GC)
>94.0%

Description

2-(2-Fluorophenyl)pyrrolidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Fluorophenyl)pyrrolidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in freezer(-20°C). Keep container tightly closed. Store in a cool, dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text