Sand, Extra Pure, SLR, Low Iron, 40-100 Mesh, Fisher Chemical™
Sand, Extra Pure, SLR, Low Iron, 40-100 Mesh, Fisher Chemical™
Sand, Extra Pure, SLR, Low Iron, 40-100 Mesh, Fisher Chemical™
Sand, Extra Pure, SLR, Low Iron, 40-100 Mesh, Fisher Chemical™
Fisher Chemical™

Sand, Extra Pure, SLR, Low Iron, 40-100 Mesh, Fisher Chemical™

Have Questions?
Catalog NumberQuantity
S-0360-63
also known as S/0360/63
3 kg
Catalog number S-0360-63
also known as S/0360/63
Price (TWD)
-
Quantity:
3 kg
Request bulk or custom format
Specifications
Boiling Point2230°C
CAS14808-60-7
ChEBICHEBI:30563
ColorBeige
Formula Weight60.08g/mol
View more
Ask our AI about this Product
Sand, Extra Pure, SLR, Low Iron, 40-100 Mesh
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection