Gadolinium(III) chloride hexahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gadolinium(III) chloride hexahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 g
5 g
25 g
1 kg
產品號碼 011286.22
亦稱為 011286-22
價格 (TWD)/ Each
-
申請報價
Quantity:
100 g
Request bulk or custom format
化學識別
CAS13450-84-5
IUPAC Namegadolinium(3+) hexahydrate trichloride
Molecular FormulaCl3GdH12O6
InChI KeyPNYPSKHTTCTAMD-UHFFFAOYSA-K
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Gd+3]
檢視更多
規格Specification Sheet規格表
Total Rare Earth Oxide Impurities0.1% max.
Gadolinium(III) chloride hexahydrate is used as a macrophage inhibitor which inhibits the specialized liver macrophages, Kupffer cells that line the walls of the sinusoids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only