Thermo Scientific Chemicals

Hafnium carbide, Gray powder, 99.5% (Metals basis excluding Zr), Thermo Scientific Chemicals

產品號碼: 012567.18
50 g, Each
Thermo Scientific Chemicals

Hafnium carbide, Gray powder, 99.5% (Metals basis excluding Zr), Thermo Scientific Chemicals

產品號碼: 012567.18
50 g, Each
Quantity
產品號碼: 012567.18
價格 (TWD)

化學識別

CAS
12069-85-1
IUPAC Name
methanidylidynehafniumylium
Molecular Formula
CHf
InChI Key
NVDNLVYQHRUYJA-UHFFFAOYSA-N
SMILES
[C-]#[Hf+]
Total Metal Impurities excluding Zr
0.5% max.
Impurity content
Zr <1%

說明

It is used as an intermediate in chemical research, Pharmaceutical Intermediates,Organic Intermediates. Used in rocket nozzle, especially for re-entry space rocket nose cone.Due to its high hardness and melting point, HfC also widely use in hard alloy additives.Used in ceramic industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text