Thermo Scientific Chemicals

Lead(II) iodide, 99.9985% (metals basis), Thermo Scientific Chemicals

產品號碼: 012724.22
100 g, Each
Thermo Scientific Chemicals

Lead(II) iodide, 99.9985% (metals basis), Thermo Scientific Chemicals

產品號碼: 012724.22
100 g, Each
Quantity
產品號碼: 012724.22
亦稱為 012724-22
價格 (TWD)

化學識別

CAS
10101-63-0
IUPAC Name
λ²-lead(2+) diiodide
Molecular Formula
I2Pb
InChI Key
RQQRAHKHDFPBMC-UHFFFAOYSA-L
SMILES
[I-].[I-].[Pb++]
Form
Powder
Appearance (Color)
Yellow
Total Metal Impurities
≤0.0015% (15 ppm)

說明

Lead(II) iodide is used as a detector material for high energy photons including x-rays and gamma rays. It is used in photography, printing, mosaic gold, and bronzing. It exhibits ferroelastic properties and has efficiency in stopping X-ray and gamma ray, which provides excellent environmental stability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text