Nitric acid, 70%, Thermo Scientific Chemicals
Nitric acid, 70%, Thermo Scientific Chemicals
Nitric acid, 70%, Thermo Scientific Chemicals
Nitric acid, 70%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nitric acid, 70%, Thermo Scientific Chemicals

May darken on storage
Have Questions?
變更視圖buttonViewtableView
Quantity:
2.5 L
1 L
產品號碼 124650025
價格 (TWD)
-
申請報價
Quantity:
2.5 L
Request bulk or custom format
化學識別
CAS7697-37-2
IUPAC Namenitric acid
Molecular FormulaHNO3
InChI KeyGRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILESO[N+]([O-])=O
檢視更多
規格Specification Sheet規格表
Titration with NaOH67 to 70 %
Appearance (Form)Clear liquid
AppearanceFree from suspended matter or sediment
Color scale=<30 APHA
AppearanceMay darken on storage
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

文件與下載

證書

安全資料表

分享目錄編號、產品名稱或連結。