Manganese(IV) oxide, 99.99+%, (trace metal basis), Thermo Scientific Chemicals
Manganese(IV) oxide, 99.99+%, (trace metal basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Manganese(IV) oxide, 99.99+%, (trace metal basis), Thermo Scientific Chemicals

Have Questions?
變更視圖buttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
產品號碼 193471000
價格 (TWD)/ Each
-
申請報價
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
IUPAC Namedioxomanganese
Molecular FormulaMnO2
InChI KeyNUJOXMJBOLGQSY-UHFFFAOYSA-N
SMILESO=[Mn]=O
Molecular Weight (g/mol)86.94
檢視更多
規格Specification Sheet規格表
Appearance (Form)Powder or crystals and/or chunks
Appearance (Color)Grey to black
Total trace metal impurities=<100 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis, Oxidation reagent
RUO – Research Use Only