化學識別
CAS2044-85-1
IUPAC Name3'-(acetyloxy)-2',7'-dichloro-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthen]-6'-yl acetate
Molecular FormulaC24H14Cl2O7
InChI KeyVQVUBYASAICPFU-UHFFFAOYSA-N
SMILESCC(=O)OC1=CC2=C(C=C1Cl)C1(OC(=O)C3=CC=CC=C13)C1=CC(Cl)=C(OC(C)=O)C=C1O2
檢視更多