Methyl 5-hexynoate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl 5-hexynoate, 95%, Thermo Scientific Chemicals
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
產品號碼 380880050
價格 (TWD)/ Each
-
申請報價
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS77758-51-1
IUPAC Namemethyl hex-5-ynoate
Molecular FormulaC7H10O2
InChI KeyLZULAZTXJLWELL-UHFFFAOYSA-N
SMILESCOC(=O)CCCC#C
檢視更多
規格Specification Sheet規格表
Refractive index1.4330 to 1.4350 (20°C, 589 nm)
GC>=94 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only