Furfuryl alcohol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Furfuryl alcohol, 98%, Thermo Scientific Chemicals
Have Questions?
變更視圖buttonViewtableView
Quantity:
1000 g
50 g
250 g
產品號碼 A10968.0B
亦稱為 A10968-0B
價格 (TWD)/ Each
-
申請報價
Quantity:
1000 g
Request bulk or custom format
化學識別
CAS98-00-0
IUPAC Name(furan-2-yl)methanol
Molecular FormulaC5H6O2
InChI KeyXPFVYQJUAUNWIW-UHFFFAOYSA-N
SMILESOCC1=CC=CO1
檢視更多
規格Specification Sheet規格表
Refractive Index1.4850-1.4890 @ 20?C
Appearance (Color)Clear colorless to yellow
Assay (GC)≥97.5%
FormLiquid
Identification (FTIR)Conforms
Furfuryl alcohol was used in the synthesis of continuous titanium carbide nanofibers/nanoribbon. Furfuryl alcohol is an efficient trapping agent for singlet oxygen determination in natural waters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Furfuryl alcohol was used in the synthesis of continuous titanium carbide nanofibers/nanoribbon. Furfuryl alcohol is an efficient trapping agent for singlet oxygen determination in natural waters.

Solubility
Soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep containers tightly closed in a cool, well-ventilated place. Keep in properly labeled containers. Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Werner R. Haag.; Jürg Hoigne.; Ernst Gassman.; André M. Braun. Singlet oxygen in surface waters — Part I: Furfuryl alcohol as a trapping agent. Chemosphere. 1984, 13, (5-6), 631-640.
  2. Mekki Choura.; Naceur M. Belgacem.; Alessandro Gandini. Acid-Catalyzed Polycondensation of Furfuryl Alcohol:  Mechanisms of Chromophore Formation and Cross-Linking. Macromolecules. 1996, 29, (11), 3839-3850.