Thermo Scientific Chemicals

3,4-(Methylenedioxy)aniline, 98+%, Thermo Scientific Chemicals

產品號碼: A12975.09
10 g, Each
Thermo Scientific Chemicals

3,4-(Methylenedioxy)aniline, 98+%, Thermo Scientific Chemicals

產品號碼: A12975.09
10 g, Each
Quantity
產品號碼: A12975.09
亦稱為 A12975-09
價格 (TWD)

化學識別

CAS
14268-66-7
IUPAC Name
2H-1,3-benzodioxol-5-amine
Molecular Formula
C7H7NO2
InChI Key
XGNXYCFREOZBOL-UHFFFAOYSA-N
SMILES
NC1=CC=C2OCOC2=C1
Appearance (Color)
Dark brown to brown to black
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
39.0-47.0?C
Form
Crystals or powder or crystalline powder or chunks or fused solid or liquid as melt

說明

3,4-(Methylenedioxy)aniline is an pharmaceutically important aniline derivative. It undergoes N-alkylation with cyclic secondary alkylamines in the presence of Shvo catalyst to yield N-arylpyrrolidines. 3,4-(Methylenedioxy)aniline was used in the synthesis of γ-glutamylanilides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-(Methylenedioxy)aniline is an pharmaceutically important aniline derivative. It undergoes N-alkylation with cyclic secondary alkylamines in the presence of Shvo catalyst to yield N-arylpyrrolidines. 3,4-(Methylenedioxy)aniline was used in the synthesis of γ-glutamylanilides.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong acids, acid chlorides, acid anhydrides. Air & Light Sensitive. Stable under recommended storage conditions.
RUO – Research Use Only

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text