2-Phenyl-2-pentenal, (E)+(Z), 90+%, Thermo Scientific Chemicals
2-Phenyl-2-pentenal, (E)+(Z), 90+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Phenyl-2-pentenal, (E)+(Z), 90+%, Thermo Scientific Chemicals

Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
產品號碼 A13635.06
亦稱為 A13635-06
價格 (TWD)/ Each
-
申請報價
Quantity:
5 g
Request bulk or custom format
化學識別
CAS3491-63-2
IUPAC Name(2E)-2-phenylpent-2-enal
Molecular FormulaC11H12O
InChI KeyYPAJRUMMODCONM-WDZFZDKYSA-N
SMILESCCC=C(C=O)C1=CC=CC=C1
檢視更多
規格Specification Sheet規格表
Assay (GC)≥90.0%
Appearance (Color)Clear colorless to yellow
Free acid (titration)≤5%
Identification (FTIR)Conforms
FormLiquid
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only