Thermo Scientific Chemicals

Methyl 3-mercaptopropionate, 98%, Thermo Scientific Chemicals

產品號碼: B20718.36
500 g, Each
Thermo Scientific Chemicals

Methyl 3-mercaptopropionate, 98%, Thermo Scientific Chemicals

產品號碼: B20718.36
500 g, Each
Quantity
產品號碼: B20718.36
亦稱為 B20718-36
價格 (TWD)

化學識別

CAS
2935-90-2
IUPAC Name
methyl 3-sulfanylpropanoate
Molecular Formula
C4H8O2S
InChI Key
LDTLDBDUBGAEDT-UHFFFAOYSA-N
SMILES
COC(=O)CCS
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.4615-1.4655 @ 20?C

說明

Methyl 3-mercaptopropionate is a click chemistry reagent used in the modification of oxazolines using thiol-ene click modification. It also has implications for self-assembled monolayers (SAMs). It is used in the colorimetric detection of mercury(II) in a high-salinity solution using gold nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text