Thermo Scientific Chemicals

4-Methylumbelliferyl-beta-D-glucuronide, 98%, Thermo Scientific Chemicals

產品號碼: B21190.MD
250 mg, Each
Thermo Scientific Chemicals

4-Methylumbelliferyl-beta-D-glucuronide, 98%, Thermo Scientific Chemicals

產品號碼: B21190.MD
250 mg, Each
Quantity
產品號碼: B21190.MD
亦稱為 B21190-MD
價格 (TWD)

化學識別

CAS
6160-80-1
IUPAC Name
(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[(4-methyl-2-oxo-2H-chromen-7-yl)oxy]oxane-2-carboxylic acid
Molecular Formula
C16H16O9
InChI Key
ARQXEQLMMNGFDU-JHZZJYKESA-N
SMILES
CC1=CC(=O)OC2=CC(O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(O)=O)=CC=C12
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Comment
Optical Rotation varies with water content
Water Content (Karl Fischer Titration)
≤20%
Optical Rotation
-105.0? ± 5.0? (c=0.1 in water)

說明

It is a fluorogenic substrate for measuring of β-glucoronidiase activity, for rapid sensitive identification of Escherichia coli. MUG is a substrate for glucuronidase. It is ideal for plant molecular study as a way to examine gene expression under various promoters. And while it works well for measuring activity in plant lysate and leaf disks, it can also work in whole plant assays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a fluorogenic substrate for measuring of ß-glucoronidiase activity, for rapid sensitive identification of Escherichia coli. MUG is a substrate for glucuronidase. It is ideal for plant molecular study as a way to examine gene expression under various promoters. And while it works well for measuring activity in plant lysate and leaf disks, it can also work in whole plant assays.

Solubility
Soluble in hot water.

Notes
Light sensitive. Store away from alkali metals bases and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in freezer at -20°C.
RUO – Research Use Only

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text