Thermo Scientific Chemicals

2,4-Difluoro-3-methoxybenzoic acid, 98+%, Thermo Scientific Chemicals

產品號碼: B25597.06
5 g, Each
Thermo Scientific Chemicals

2,4-Difluoro-3-methoxybenzoic acid, 98+%, Thermo Scientific Chemicals

產品號碼: B25597.06
5 g, Each
Quantity
產品號碼: B25597.06
亦稱為 B25597-06
價格 (TWD)
Have Questions?

化學識別

CAS
178974-97-5
Assay (HPLC)
≥98.0%
Melting Point (clear melt)
191.0-197.0?C
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%

說明

2,4-Difluoro-3-methoxybenzoic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Difluoro-3-methoxybenzoic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text