Thermo Scientific Chemicals

N-Boc-4-iodo-L-phenylalanine, 98%, Thermo Scientific Chemicals

產品號碼: H51960.06
5 g, Each
Thermo Scientific Chemicals

N-Boc-4-iodo-L-phenylalanine, 98%, Thermo Scientific Chemicals

產品號碼: H51960.06
5 g, Each
Quantity
產品號碼: H51960.06
亦稱為 H51960-06
價格 (TWD)

化學識別

CAS
62129-44-6
IUPAC Name
(2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-iodophenyl)propanoic acid
Molecular Formula
C14H18INO4
InChI Key
JZLZDBGQWRBTHN-NSHDSACASA-N
SMILES
CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(I)C=C1)C(O)=O
Assay (HPLC)
97.5% min

說明

N-Boc-4-iodo-L-phenylalanine is used as pharmaceutical intermediate. It is also used as a p-iodo phenyl alanine derivative with N-Boc protection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-4-iodo-L-phenylalanine is used as pharmaceutical intermediate. It is also used as a p-iodo phenyl alanine derivative with N-Boc protection.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text