Thermo Scientific Chemicals

N-Boc-4-methyl-L-phenylalanine, 95%, Thermo Scientific Chemicals

產品號碼: H51983.06
5 g, Each
Thermo Scientific Chemicals

N-Boc-4-methyl-L-phenylalanine, 95%, Thermo Scientific Chemicals

產品號碼: H51983.06
5 g, Each
Quantity
產品號碼: H51983.06
亦稱為 H51983-06
價格 (TWD)
Have Questions?

說明

It is used as an active pharmaceutical intermediate. It is also an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate. It is also an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text