Thermo Scientific Chemicals

3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate, 98+%, Thermo Scientific Chemicals

產品號碼: H56061.18
50 g, Each
Thermo Scientific Chemicals

3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate, 98+%, Thermo Scientific Chemicals

產品號碼: H56061.18
50 g, Each
Quantity
產品號碼: H56061.18
亦稱為 H56061-18
價格 (TWD)

化學識別

CAS
38894-11-0
IUPAC Name
(2Z)-2-hydrazinylidene-3-methyl-2,3-dihydro-1,3-benzothiazole
Molecular Formula
C8H9N3S
InChI Key
PHOLIFLKGONSGY-NTMALXAHSA-N
SMILES
CN1C(SC2=CC=CC=C12)=NN
Assay (HPLC)
>98.0%

說明

It is used as a reagent for the spectrophotometric determination of aliphatic aldehydes and for the determination of hexosamines in glycosaminoglycans. It is Incorporated into a new peroxidase color reaction. It is also a reagent for the spectrophotometric determination of traces of selenium (IV) in environmental samples.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a reagent for the spectrophotometric determination of aliphatic aldehydes and for the determination of hexosamines in glycosaminoglycans. It is Incorporated into a new peroxidase color reaction. It is also a reagent for the spectrophotometric determination of traces of selenium (IV) in environmental samples.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents. Store at room temperature.
RUO – Research Use Only

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text