Thermo Scientific Chemicals

N-Boc-N-methyl-L-serine, 98%, Thermo Scientific Chemicals

產品號碼: H63314.06
5 g, Each
Thermo Scientific Chemicals

N-Boc-N-methyl-L-serine, 98%, Thermo Scientific Chemicals

產品號碼: H63314.06
5 g, Each
Quantity
產品號碼: H63314.06
亦稱為 H63314-06
價格 (TWD)

化學識別

CAS
101772-29-6
IUPAC Name
2-{[(tert-butoxy)carbonyl](methyl)amino}-3-hydroxypropanoic acid
Molecular Formula
C9H17NO5
InChI Key
NJQGIDVCNBZXLG-UHFFFAOYNA-N
SMILES
CN(C(CO)C(O)=O)C(=O)OC(C)(C)C
Appearance (Color)
White
Form
Solid
Assay (unspecified)
>96.0% (NMR)

說明

N-Boc-N-methyl-L-serine is used as a precursor in organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-N-methyl-L-serine is used as a precursor in organic synthesis and pharmaceuticals.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text