Thermo Scientific Chemicals

Sotalol hydrochloride, 98%, Thermo Scientific Chemicals

產品號碼: J63772.03
1 g, Each
Thermo Scientific Chemicals

Sotalol hydrochloride, 98%, Thermo Scientific Chemicals

產品號碼: J63772.03
1 g, Each
Quantity
產品號碼: J63772.03
亦稱為 J63772-03
價格 (TWD)

化學識別

CAS
959-24-0
Assay (unspecified)
99.0 - 101.0% (Calculated on dry basis)
Form
White to pale yellow
Loss on Drying
≤0.5%

說明

Sotalol hydrochloride is used as a potent beta adrenergic antagonist, prolongs the action potential and increases the refractory period. Sotalol hydrochloride is also considered a non-selective β blocker and a potassium channel blocker with an IC50 of 43 μM.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sotalol hydrochloride is used as a potent beta adrenergic antagonist, prolongs the action potential and increases the refractory period. Sotalol hydrochloride is also considered a non-selective β blocker and a potassium channel blocker with an IC50 of 43 µM.

Solubility
Soluble in phosphate buffered saline, DMSO, ethanol, water, and methanol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text