Thermo Scientific Chemicals

2,3-O-Isopropylidene-D-ribonic acid-1,4-lactone, 97+%, Thermo Scientific Chemicals

產品號碼: J66442.ME
500 mg, Each
Thermo Scientific Chemicals

2,3-O-Isopropylidene-D-ribonic acid-1,4-lactone, 97+%, Thermo Scientific Chemicals

產品號碼: J66442.ME
500 mg, Each
Quantity
產品號碼: J66442.ME
亦稱為 J66442-ME
價格 (TWD)

化學識別

CAS
30725-00-9
IUPAC Name
6-(hydroxymethyl)-2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-one
Molecular Formula
C8H12O5
InChI Key
NHHKFJCWLPPNCN-UHFFFAOYNA-N
SMILES
CC1(C)OC2C(CO)OC(=O)C2O1
Assay (unspecified)
Specific rotation: -65? to -71?
Assay (HPLC)
>97.5%

說明

2,3-O-Isopropylidene-D-ribonic acid-1,4-lactone is used for the synthesis of C-nucleosides, prostanoids and GABA analogs via homochiral 2-piperidones. It is the starting material for the preparation of biochemical compounds including C-nucleosides, such as neplanocin A, and GABA analogs, via homochiral 2-piperidones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-O-Isopropylidene-D-ribonic acid-1,4-lactone is used for the synthesis of C-nucleosides, prostanoids and GABA analogs via homochiral 2-piperidones. It is the starting material for the preparation of biochemical compounds including C-nucleosides, such as neplanocin A, and GABA analogs, via homochiral 2-piperidones.

Solubility
Soluble in methanol at 50mg/ml, acetone, dichloromethane, ethyl Acetate.

Notes
Soluble in methanol at 50mg/ml
RUO – Research Use Only

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text