Thermo Scientific Chemicals

2-(Methylsulfonyl)ethanol, 97%, Thermo Scientific Chemicals

產品號碼: L03159.06
5 g, Each
Thermo Scientific Chemicals

2-(Methylsulfonyl)ethanol, 97%, Thermo Scientific Chemicals

產品號碼: L03159.06
5 g, Each
Quantity
產品號碼: L03159.06
亦稱為 L03159-06
價格 (TWD)

化學識別

CAS
15205-66-0
IUPAC Name
2-methanesulfonylethan-1-ol
Molecular Formula
C3H8O3S
InChI Key
KFTYFTKODBWKOU-UHFFFAOYSA-N
SMILES
CS(=O)(=O)CCO
Appearance (Color)
Clear, colorless to pale yellow to pale grey
Form
Liquid as melt
Assay (Silylated GC)
≥96.0% (UK-sourced material)
Assay from Supplier's CofA
≥96.0% (US-sourced material)
Comment
Material sourced in UK and US

說明

2-(Methylsulfonyl)ethanol may be used as reagent in the synthesis of phenols from aryl fluorides. It may be used in the preparation of 2-(methylsulfonyl)ethyl N,N-bis(2-chloroethyl)phosphorodiamidate. 2-Hydroxyethyl Methyl Sulfone, is the versatile building block used in the synthesis of more complex sulfur containing compounds. It can be used for the studies of inhibitor and substrate moieties to acetylcholinesterase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylsulfonyl)ethanol may be used as reagent in the synthesis of phenols from aryl fluorides. It may be used in the preparation of 2-(methylsulfonyl)ethyl N,N-bis(2-chloroethyl)phosphorodiamidate. 2-Hydroxyethyl Methyl Sulfone,
RUO – Research Use Only

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text