Thermo Scientific Chemicals

4-Bromo-1,1,2-trifluoro-1-butene, 98%, Thermo Scientific Chemicals

產品號碼: L12766.09
10 g, Each
Thermo Scientific Chemicals

4-Bromo-1,1,2-trifluoro-1-butene, 98%, Thermo Scientific Chemicals

產品號碼: L12766.09
10 g, Each
Quantity
產品號碼: L12766.09
亦稱為 L12766-09
價格 (TWD)

說明

4-Bromo-1,1,2-trifluoro-1-butene is used in organic synthesis, chemical research and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-1,1,2-trifluoro-1-butene is used in organic synthesis, chemical research and pharmaceuticals.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
RUO – Research Use Only

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text