Thermo Scientific Chemicals

1,1,3,3,5,5-Hexamethyltrisiloxane, 95%, Thermo Scientific Chemicals

產品號碼: L16520.14
25 g, Each
Thermo Scientific Chemicals

1,1,3,3,5,5-Hexamethyltrisiloxane, 95%, Thermo Scientific Chemicals

產品號碼: L16520.14
25 g, Each
Quantity
產品號碼: L16520.14
亦稱為 L16520-14
價格 (TWD)

化學識別

CAS
1189-93-1
Molecular Formula
C6H18O2Si3
InChI Key
YTEISYFNYGDBRV-UHFFFAOYSA-N
SMILES
C[Si](C)O[Si](C)(C)O[Si](C)C
Molecular Weight (g/mol)
206.46
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥94.0%
Refractive Index
1.3785-1.3835 @ 20?C

說明

1,1,3,3,5,5-Hexamethyltrisiloxane is involved in the preparation of 2-bis(3-allyl-4-hydroxyphenyl)hexafluoropropane, which is associated with inorganic siloxane polymer and used as a coating material for the detection of toxic chemical warfare agents. It is also used as a silylating agent and to prepare 1,5-Bis[2-(1,2-epoxycyclohex-4-yl)ethyl]-1,1,3,3,5,5-hexamethyltrisiloxane from 4-vinyl-1-cyclohexene-1,2-epoxide by hydrosilylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,3,3,5,5-Hexamethyltrisiloxane is involved in the preparation of 2-bis(3-allyl-4-hydroxyphenyl)hexafluoropropane, which is associated with inorganic siloxane polymer and used as a coating material for the detection of toxic chemical warfare agents. It is also used as a silylating agent and to prepare 1,5-Bis[2-(1,2-epoxycyclohex-4-yl)ethyl]-1,1,3,3,5,5-hexamethyltrisiloxane from 4-vinyl-1-cyclohexene-1,2-epoxide by hydrosilylation.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text