Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Proteins and Peptides
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Gene Expression
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins and Peptides
  • ELISA Kits
  • SNP Genotyping
  • 所有文檔和支持
  • 證書
  • SDS(安全資料表
  • 產品常見問題
  • Documents & Support
  • 手冊和試驗方法
    Search button Close
            • 訂單查詢
            • 快速訂購
            • 登入
              登入
              還沒有帳戶嗎 ? 立即註冊
              • 帳戶
              • 訂單
              • Connect: Lab, Data, Apps
              • 客製化產品與項目
              • 儀器管理

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Proteins & Peptides ›
            • SARS-CoV-2 Spike Protein S1 Proteins

            Invitrogen

            SARS-CoV-2 Spike Protein S1 (AY.4.2) His-tag Recombinant Protein

            View all (87) SARS-CoV-2 Spike Protein S1 proteins
            Datasheet
            Tech Support
            Datasheet
            Tech Support

            Cite SARS-CoV-2 Spike Protein S1 (AY.4.2) His-tag Recombinant Protein

            • Testing Data (1)
            Application
            SARS-CoV-2 Spike Protein S1 (AY.4.2) His-tag Protein in SDS-PAGE (SDS-PAGE)
            Group 53 Created with Sketch.

            FIGURE: 1 / 1

            SARS-CoV-2 Spike Protein S1 (AY.4.2) His-tag Protein (RP-88524) in SDS-PAGE

            SDS-PAGE analysis of SARS-CoV-2 Spike Protein S1 (AY.4.2) His-tag (Product # RP-88524). The specification for purity shows: > 95 % (SDS-PAGE), > 95 % (SEC-HPLC).
            View Product

            SARS-CoV-2 Spike Protein S1 (AY.4.2) His-tag Protein in SDS-PAGE (SDS-PAGE)

            Product Details

            RP-88524

            Applications
            Tested Dilution
            Publications

            Control (Ctrl)

            Assay-dependent
            -
            Product Specifications

            Species

            Virus

            Expression System

            HEK293 cells

            Amino acid sequence

            SARS-CoV-2 spike S1 (Accession # YP_009724390.1), amino acids Met1-Arg685 with T19R, T95I, G142D, Y145H, E156G, 157-158 deletion, A222V, L452R, T478K, D614G, P681R substitutions and a C-terminal His-tag

            Tag

            His-tag

            Molecular weight

            102.3 kDa

            Class

            Recombinant

            Type

            Protein

            Purity

            >95% as determined by SDS-PAGE. >95% as determined by SEC-HPLC.

            Endotoxin concentration

            <1.0 EU/µg

            Conjugate

            Unconjugated

            Form

            Lyophilized

            Purification

            purified

            Storage buffer

            PBS, pH 7.4, with 0.01% mannitol, 5-8% trehalose, 0.01% Tween 80

            Contains

            no preservative

            Storage conditions

            -20°C, Avoid Freeze/Thaw Cycles

            Shipping conditions

            Wet ice

            Product Specific Information

            It is recommended that the protein be aliquoted for optimal storage.

            The mutations were identified in the SARS-CoV-2 varient known as AY.4.2

            Target Information

            SARS-CoV-2 (Severe Acute Respiratory Syndrome Coronavirus 2) also known as 2019-nCoV (2019 Novel Coronavirus) is a virus that causes illnesses ranging from the common cold symptoms to severe effects including death and respiratory distress. SARS-CoV-2 Spike Protein is composed of S1 domain and S2 domain (Spike Protein). The Spike protein is the main surface antigen of the coronavirus. The glycosylated Spike protein (as well as the nucleocapsid protein) can be detected in infected cell culture supernatants with antisera from SARS patients. S1 contains a receptor-binding domain (RBD) that can specifically bind to angiotensin-converting enzyme 2 (ACE2), the receptor on target cells. CoV diversity is reflected in the variable Spike proteins (S proteins), which have evolved into forms differing in their receptor interactions and their response to various environmental triggers of virus-cell membrane fusion.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product
            It has to be done as per old AB suggested Products section.
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            產品訂購 Plus Icon Minus Icon
            • 訂單狀態
            • 訂購協助
            • 快速訂購
            • 現貨供應中心
            • 電子採購
            支持 Plus Icon Minus Icon
            • 協助與支持
            • 聯絡我們
            • 技術支持中心
            • 搜尋文件和證書
            • 回報網站問題
            資源 Plus Icon Minus Icon
            • 學習中心
            • 促銷活動
            • 活動和網路研討會
            • 社群媒體
            關於 Thermo Fisher Plus Icon Minus Icon
            • 關於我們 關於我們
            • 人才招募 人才招募
            • 投資者資訊 投資者資訊
            • 新聞 新聞
            • 企業社會責任 企業社會責任
            • 商標
            品牌 Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Policy
            • Price & Freight Policy
            • Cookie偏好

              在此網站使用 Cookie 的選擇

              我們以及我們的附屬公司和供應商使用 cookies 和類似技術來經營我們的網站、識別我們網站的訪客、提供安全登入、收集統計資料以優化網站功能,並提供根據您的興趣量身定制的內容。點擊「全部接受」 以接受所有 cookies,並直接進入網站,或點擊「管理設定」 以查看 cookies 類型的詳細說明,並選擇是否在網站接受某些 cookies。
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            Taiwan flag icon
            Taiwan

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-mfpfg:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0