Gene Transcripts

Gene Symbol
CIB3
Entrez Gene ID
117286
Gene Name
calcium and integrin binding family member 3
Gene Aliases
KIP3
Chromosome Location
Chr. 19: 16161368 - 16173525 on Build GRCh38
UniGene ID
Hs.255432
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_054113.3 NP_473454.1 2, 3 132
GenBank mRNA AB050868.1 - 2, 3 82
BC112200.1 - 2, 3 132 8327668
BC113591.1 - 2, 3 132 8322643

View Full Product Details