Gene Transcripts

Gene Symbol
Cib3
Entrez Gene ID
685723
Gene Name
calcium and integrin binding family member 3
Gene Aliases
Chromosome Location
Chr. 16: 19323516 - 19333361 on Build Rnor_6.0
UniGene ID
Rn.228176
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191081.1 NP_001178010.1 4 232
XM_008771148.1 XP_008769370.1 2 85

View Full Product Details