Gene Transcripts

Gene Symbol
Cib3
Entrez Gene ID
234421
Gene Name
calcium and integrin binding family member 3
Gene Aliases
C730014M21Rik, Gm1107, KIP3
Chromosome Location
Chr. 8: 72192699 - 72213092 on Build GRCm38
UniGene ID
Mm.211247
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080812.1 NP_001074281.1 3 220
GenBank mRNA AK050094.1 - 3 220
BB870365.1 - 3 245
BC120812.1 - 3 202 40129735
BC120814.1 - 3 202 40129737

View Full Product Details