Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_000207.2 | NP_000198.1 | ||||
NM_001185097.1 | NP_001172026.1 | |||||
NM_001185098.1 | NP_001172027.1 | |||||
NM_001291897.1 | NP_001278826.1 | |||||
GenBank mRNA | AY899304.1 | - | ||||
BC005255.1 | - | 3950204 | ||||
BM510347.1 | - | |||||
BM510748.1 | - | |||||
BP322143.1 | - | |||||
BT006808.1 | - | |||||
DQ778082.1 | - | |||||
ENST00000250971.0 | - | |||||
JF909299.1 | - | |||||
JQ951950.1 | - | |||||
X70508.1 | - |