Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001291428.1 | NP_001278357.1 | ||||
NM_001291429.1 | NP_001278358.1 | |||||
NM_001291430.1 | NP_001278359.1 | |||||
NM_001291431.1 | NP_001278360.1 | |||||
NM_004324.3 | NP_004315.1 | |||||
NM_138761.3 | NP_620116.1 | |||||
NM_138763.3 | NP_620118.1 | |||||
NM_138764.4 | NP_620119.2 | |||||
NR_027882.1 | ||||||
XM_017027077.1 | XP_016882566.1 | |||||
GenBank mRNA | AF007826.1 | - | ||||
AF247393.1 | - | |||||
AF250190.1 | - | |||||
AI565203.1 | - | |||||
AJ417988.1 | - | |||||
AJ586909.1 | - | |||||
AJ586910.1 | - | |||||
AK291076.1 | - | |||||
BC014175.2 | - | 4578562 | ||||
BE396495.1 | - | |||||
BM673184.1 | - | |||||
BM706954.1 | - | |||||
CN281003.1 | - | |||||
ENST00000293288.0 | - | |||||
HQ889830.1 | - | |||||
JX524562.1 | - | |||||
KF830846.1 | - | |||||
KF830847.1 | - | |||||
KF830848.1 | - | |||||
L22473.1 | - | |||||
L22474.1 | - | |||||
L22475.1 | - | |||||
T29044.1 | - | |||||
U19599.1 | - |