Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_004864.2 | NP_004855.2 | ||||
GenBank mRNA | AB000584.1 | - | ||||
AF003934.1 | - | |||||
AF019770.1 | - | |||||
AF173860.1 | - | |||||
AK225431.1 | - | |||||
AK291530.1 | - | |||||
BC000529.2 | - | 2961614 | ||||
BC008962.2 | - | 3009858 | ||||
BT019465.1 | - | |||||
ENST00000252809.0 | - | |||||
U51731.1 | - | |||||
U88323.1 | - |