Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001278736.1 | NP_001265665.1 | ||||
NM_002985.2 | NP_002976.2 | |||||
GenBank mRNA | AA443491.1 | - | ||||
AA486131.1 | - | |||||
AF043341.1 | - | |||||
AF266753.1 | - | |||||
AK312212.1 | - | |||||
AW769950.1 | - | |||||
BC008600.1 | - | 4185200 | ||||
BG272739.1 | - | |||||
CB111656.1 | - | |||||
CD695497.1 | - | |||||
CR980938.1 | - | |||||
DQ230537.1 | - | |||||
GD136908.1 | - | |||||
HY265548.1 | - | |||||
M21121.1 | - |