Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_000215.3 | NP_000206.2 | ||||
XM_005259896.3 | XP_005259953.1 | |||||
XM_011527990.1 | XP_011526292.1 | |||||
XM_011527991.2 | XP_011526293.1 | |||||
GenBank mRNA | AK296383.1 | - | ||||
AK304392.1 | - | |||||
BC028068.1 | - | 5212575 | ||||
BF512748.1 | - | |||||
BM561964.1 | - | |||||
DA405504.1 | - | |||||
ENST00000458235.0 | - | |||||
KU178153.1 | - | |||||
KU178154.1 | - | |||||
U08340.1 | - | |||||
U09607.1 | - | |||||
U31317.1 | - | |||||
U31601.1 | - | |||||
U57096.1 | - |